6-Hydroxy-5-nitronicotinic acid ≥98%

Supplier: Matrix Scientific

026730-5G 026730-25G
101822-744EA 36.92 USD
101822-744 101822-746
6-Hydroxy-5-nitronicotinic acid ≥98%
6-Hydroxy-5-nitronicotinic acid

Hazard: Irritant

Formula: C₆H₄N₂O₅
MW: 184.1100 g/mol
Melting Pt: 269-271°C
MDL Number: MFCD02180840
CAS Number: 6635-31-0

Order Now

Specification Test Results

Melting point 269-271°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR