3-(Trifluoromethoxy)benzenesulfonamide ≥97%

Supplier: Matrix Scientific

026308-1G 026308-5G
101822-052EA 60 USD
101822-052 101822-054
3-(Trifluoromethoxy)benzenesulfonamide ≥97%
3-(Trifluoromethoxy)benzenesulphonamide

Hazard: Irritant

Formula: C₇H₆F₃NO₃S
MW: 241.19 g/mol
Melting Pt: 100…103 °C
MDL Number: MFCD03094233
CAS Number: 503179-70-2

Order Now

Specification Test Results

Melting point 100-103°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR