4-(4-Fluorophenyl)butyric acid ≥97%

Supplier: Matrix Scientific

020616-1G 020616-5G
101810-462EA 110.77 USD
101810-462 101810-464
4-(4-Fluorophenyl)butyric acid ≥97%
4-(4-Fluorophenyl)butyric acid

Hazard: Irritant

Formula: C₁₀H₁₁FO₂
Melting Pt: 45…47 °C
MDL Number: MFCD03788503
CAS Number: 589-06-0

Order Now

Specification Test Results

Melting point 45-47°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR