4-Chloro-2,5-xylidine ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-2,5-dimethylaniline

020612-1G 020612-5G
101810-442EA 21.54 USD
101810-442 101810-444
4-Chloro-2,5-xylidine ≥98%
4-Chloro-2,5-xylidine

Hazard: Irritant

Formula: C₈H₁₀ClN
MW: 155.63 g/mol
Melting Pt: 94-96°C
MDL Number: MFCD08445705
CAS Number: 20782-94-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 94-96°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR