Benzyl(4-oxocyclohexyl)carbamate ≥97%

Supplier: Matrix Scientific

Synonyms: Benzyl (4-oxocyclohexyl)carbamate, 4-N-Cbz-aminocyclohexanone

020566-1G 020566-5G
101810-300EA 21.54 USD
101810-300 101810-302
Benzyl(4-oxocyclohexyl)carbamate ≥97%
Benzyl(4-oxocyclohexyl)carbamate

Hazard: Irritant

Formula: C₁₄H₁₇NO₃
MW: 247.29 g/mol
Melting Pt: 86…87 °C
MDL Number: MFCD06658393
CAS Number: 16801-63-1

Order Now

Specification Test Results

Melting point 86-87°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR