2-Bromo-6-nitro-4-(trifluoromethoxy)aniline ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-4-(trifluoromethoxy)-6-nitroaniline

020559-5G 020559-25G
101810-270EA 158.46 USD
101810-270 101810-272
2-Bromo-6-nitro-4-(trifluoromethoxy)aniline ≥98%
2-Bromo-6-nitro-4-(trifluoromethoxy)aniline

Hazard: Irritant

Formula: C₇H₄BrF₃N₂O₃
MW: 301.0200 g/mol
Melting Pt: 62…64 °C
MDL Number: MFCD04039212
CAS Number: 886499-21-4

Order Now

Specification Test Results

Melting point 62-64°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR