5-Bromo-3-nitropyridine-2-carbonitrile ≥97%

Supplier: Matrix Scientific

020189-5G 020189-25G
101809-578EA 20 USD
101809-578 101809-580
5-Bromo-3-nitropyridine-2-carbonitrile ≥97%
5-Bromo-3-nitropyridine-2-carbonitrile

Hazard: Irritant

Formula: C₆H₂BrN₃O₂
Melting Pt: 101…106 °C
MDL Number: MFCD06657551
CAS Number: 573675-25-9

Order Now

Specification Test Results

Melting point 101-106°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR