3,5-Dibromobenzonitrile ≥98%

Supplier: Matrix Scientific

020187-5G 020187-25G
101809-570EA 26.15 USD
101809-570 101809-572
3,5-Dibromobenzonitrile ≥98%
3,5-Dibromobenzonitrile

Hazard: Irritant

Formula: C₇H₃Br₂N
MW: 260.9134 g/mol
Melting Pt: 98°C
MDL Number: MFCD00178770
CAS Number: 97165-77-0

Order Now

Specification Test Results

Melting point 98°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR