5-Bromo-2-chloroaniline ≥98%
Supplier: Matrix Scientific
020186-25G
020186-100G
101809-566EA
73.85
USD
101809-566
101809-568
5-Bromo-2-chloroaniline ≥98%
5-Bromo-2-chloroaniline
Hazard: Irritant
Formula:
C₆H₅BrClN MW: 206.47 g/mol Melting Pt: 48-49°C |
MDL Number:
MFCD06796283 CAS Number: 60811-17-8 |
Specification Test Results
Melting point | 48-49°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...