4,4-Difluorocyclohexanone ≥97%

Supplier: Matrix Scientific

020185-1G 020185-5G
101809-560EA 46.15 USD
101809-560 101809-562
4,4-Difluorocyclohexanone ≥97%
4,4-Difluorocyclohexanone

Hazard: Irritant

Formula: C₆H₈F₂O
Melting Pt: 35…36 °C
MDL Number: MFCD08146635
CAS Number: 22515-18-0

Order Now

Specification Test Results

Melting point 35-36°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR