N-Acetyl-DL-alanine ≥98%

Supplier: Matrix Scientific

Synonyms: N-Acetyl-DL-2-aminopropionic Acid, Ac-DL-Ala-OH, N-Ac-DL-Ala-OH, N-Acetyl-DL-2-aminopropionic acid, N-Acetyl-DL-Alanine, 2-Acetylamino-propionic acid

020029-25G 020029-100G
101809-278EA 27.69 USD
101809-278 101809-280
N-Acetyl-DL-alanine ≥98%
N-Acetyl-DL-alanine

Hazard: Irritant/Store in Freezer

Formula: C₅H₉NO₃
MW: 131.13 g/mol
Melting Pt: 135…139 °C
MDL Number: MFCD00037238
CAS Number: 1115-69-1

Order Now

Specification Test Results

Melting point 135-139°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR