2-(1H-Pyrazol-4-yl)quinoxaline ≥98%

Supplier: Matrix Scientific

016113-1G 016113-5G
101804-124EA 372.31 USD
101804-124 101804-126
2-(1H-Pyrazol-4-yl)quinoxaline ≥98%
2-(1H-Pyrazol-4-yl)quinoxaline

Hazard: Irritant

Formula: C₁₁H₈N₄
Melting Pt: 204…207 °C
MDL Number: MFCD02579323
CAS Number: 439106-90-8

Order Now

Specification Test Results

Melting point 204-207°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR