4-(1H-Pyrazol-4-yl)pyrimidine ≥98%

Supplier: Matrix Scientific

016110-1G 016110-5G
101804-114EA 63.08 USD
101804-114 101804-116
4-(1H-Pyrazol-4-yl)pyrimidine ≥98%
4-(1H-Pyrazol-4-yl)pyrimidine

Hazard: Irritant

Formula: C₇H₆N₄
Melting Pt: 205…208 °C
MDL Number: MFCD02579319
CAS Number: 28648-87-5

Order Now

Specification Test Results

Melting point 205-208°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR