2-Methylsulfanyl-7H-pyrrolo[2,3-d]pyrimidin-4-ol ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Hydroxy-2-(methylthio)-7H-pyrrolo[2,3-d]pyrimidine

016104-1G 016104-5G
101804-098EA 193.85 USD
101804-098 101804-100
2-Methylsulfanyl-7H-pyrrolo[2,3-d]pyrimidin-4-ol ≥97%
2-Methylsulfanyl-7H-pyrrolo[2,3-d]pyrimidin-4-ol

Hazard: Irritant

Formula: C₇H₇N₃OS
MW: 181.22 g/mol
Melting Pt: 296…298 °C (dec.)
MDL Number: MFCD02668244
CAS Number: 67831-83-8

Order Now

Specification Test Results

Melting point 296-298°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR