3-(Benzyldimethylammonio)propanesulfonate ≥98%

Supplier: Matrix Scientific

Synonyms: NDSB-256, 3-(Benzyldimethylammonio) propane-1-sulphonate, 3-[dimethylbenzylamino]propanesulphonic acid

016085-1G 016085-5G
101804-060EA 63.08 USD
101804-060 101804-062
3-(Benzyldimethylammonio)propanesulfonate ≥98%
3-(Benzyldimethylammonio)propanesulfonate

Hazard: Irritant

Formula: C₁₂H₁₉NO₃S
MW: 257.348 g/mol
Melting Pt: 245…248 °C
MDL Number: MFCD00225018
CAS Number: 81239-45-4

Order Now

Specification Test Results

Melting point 245-248°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR