3,4,5-Trimethoxyphenylglyoxal monohydrate ≥97%

Supplier: Matrix Scientific

016082-1G 016082-5G
101804-050EA 315.38 USD
101804-050 101804-052
3,4,5-Trimethoxyphenylglyoxal monohydrate ≥97%
3,4,5-Trimethoxyphenylglyoxal monohydrate

Hazard: Irritant

Formula: C₁₁H₁₄O₆
MW: 242.23 g/mol
Melting Pt: 135 °C
MDL Number: MFCD06801328
CAS Number: 858255-78-4

Order Now

Specification Test Results

Melting point 135°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR