2-(Trifluoromethyl)phenylglyoxal hydrate ≥97%

Supplier: Matrix Scientific

016080-250MG 016080-1G
101804-040EA 516.92 USD
101804-040 101804-042
2-(Trifluoromethyl)phenylglyoxal hydrate ≥97%
2-(Trifluoromethyl)phenylglyoxal hydrate

Hazard: Irritant

Formula: C₉H₇F₃O₃
MW: 220.15 g/mol
Melting Pt: 39…42 °C
MDL Number: MFCD06801323
CAS Number: 745783-91-9

Order Now

Specification Test Results

Melting point 39-42°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR