4-Methoxyphenylglyoxal hydrate ≥97%

Supplier: Matrix Scientific

016078-1G 016078-5G
101804-032EA 76.92 USD
101804-032 101804-034
4-Methoxyphenylglyoxal hydrate ≥97%
4-Methoxyphenylglyoxal hydrate

Hazard: Irritant

Formula: C₉H₁₀O₄
Melting Pt: 112…115 °C
MDL Number: MFCD00234534
CAS Number: 16208-17-6

Order Now

Specification Test Results

Melting point 112-115°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR