3-Fluorophenylglyoxal hydrate ≥97%

Supplier: Matrix Scientific

016075-1G 016075-5G
101804-022EA 73.85 USD
101804-022 101804-024
3-Fluorophenylglyoxal hydrate ≥97%
3-Fluorophenylglyoxal hydrate

Hazard: Irritant

Formula: C₈H₇O₃F
Melting Pt: 76…78 °C
MDL Number: MFCD06800769
CAS Number: 121247-01-6

Order Now

Specification Test Results

Melting point 76-78°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR