3,4-Difluorophenylglyoxal hydrate ≥97%

Supplier: Matrix Scientific

016073-1G 016073-5G
101804-014EA 38.46 USD
101804-014 101804-016
3,4-Difluorophenylglyoxal hydrate ≥97%
3,4-Difluorophenylglyoxal hydrate

Hazard: Irritant

Formula: C8H6F2O3
Melting Pt: 78…80 °C
MDL Number: MFCD03453081
CAS Number: 1049753-77-6

Order Now

Specification Test Results

Melting point 78-80°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR