Ethyl-2,4-dihydroxy-6-methyl-3-pyridinecarboxylate ≥98%

Supplier: Matrix Scientific

Synonyms: Ethyl 2,4-Dihydroxy-6-methyl-3-pyridinecarboxylate, 2,4-Dihydroxy-6-methyl-3-pyridinecarboxylic acid ethyl ester, Ethyl 2,4-dihydroxy-6-methyl-3-pyridinecarboxylate, Ethyl 2,4-Dihydroxy-6-methylnicotinate, 2,4-Dihydroxy-6-methylnicotinic acid ethyl ester

016035-5G 016035-25G
101803-958EA 27.69 USD
101803-958 101803-960
Ethyl-2,4-dihydroxy-6-methyl-3-pyridinecarboxylate ≥98%
Ethyl-2,4-dihydroxy-6-methyl-3-pyridinecarboxylate

Hazard: Irritant

Formula: C₉H₁₁NO₄
MW: 197.1900 g/mol
Melting Pt: 205…208 °C
MDL Number: MFCD02929397
CAS Number: 70254-52-3

Order Now

Specification Test Results

Melting point 205-208°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR