1-Benzhydrylazetidin-3-ol ≥97%

Supplier: Matrix Scientific

012824-5G 012824-25G 012824-100G
101803-464EA 15.38 USD
101803-464 101803-466 102663-588
1-Benzhydrylazetidin-3-ol ≥97%
1-Benzhydrylazetidin-3-ol

Hazard: Irritant

Formula: C₁₆H₁₇NO
Melting Pt: 115 °C
MDL Number: MFCD00205109
CAS Number: 18621-17-5

Order Now

Specification Test Results

Melting point 110-113°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR