2,6-Diacetylpyridine ≥97%

Supplier: Matrix Scientific

012659-1G 012659-5G
101803-130EA 23.08 USD
101803-130 101803-132
2,6-Diacetylpyridine ≥97%
2,6-Diacetylpyridine

Hazard: Irritant

Formula: C₉H₉NO₂
Melting Pt: 80-82°C (sub)
MDL Number: MFCD00006304
CAS Number: 1129-30-2

Order Now

Specification Test Results

Melting point 80-82°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR