5-Methoxy-1H-indole-3-carbaldehyde ≥98%
Supplier: Matrix Scientific
012639-5G
012639-25G
101803-064EA
29.23
USD
101803-064
102550-974
5-Methoxy-1H-indole-3-carbaldehyde ≥98%
5-Methoxy-1H-indole-3-carbaldehyde
Hazard: Irritant/Air Sensitive/Store in Freezer
Formula:
C₁₀H₉NO₂ MW: 175.19 g/mol Melting Pt: 179…183 °C |
MDL Number:
MFCD00005623 CAS Number: 10601-19-1 |
Specification Test Results
Melting point | 179-183°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...