5-Benzyloxyindole-3-carbaldehyde ≥98%

Supplier: Matrix Scientific

Synonyms: 5-(Benzyloxy)-1H-indole-3-carbaldehyde, 5-benzyloxy-1h-indole-3-carbaldehyde, 5-Benzyloxyindole-3-carboxaldehyde

012638-5G 012638-25G
101803-058EA 20 USD
101803-058 101803-060
5-Benzyloxyindole-3-carbaldehyde ≥98%
5-Benzyloxyindole-3-carbaldehyde

Hazard: Irritant/Air Sensitive/Store in Freezer

Formula: C₁₆H₁₃NO₂
MW: 251.285 g/mol
Melting Pt: 237…238 °C
MDL Number: MFCD00014562
CAS Number: 6953-22-6

Order Now

Specification Test Results

Melting point 237-238°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR