3,5-Dibenzyloxybenzyl bromide ≥98%

Supplier: Matrix Scientific

012586-1G 012586-5G
101802-920EA 253.85 USD
101802-920 101802-922
3,5-Dibenzyloxybenzyl bromide ≥98%
3,5-Dibenzyloxybenzyl bromide

Hazard: Irritant/Refrigerate

Formula: C₂₁H₁₉BrO₂
Melting Pt: 86-88°C
MDL Number: MFCD02093444
CAS Number: 24131-32-6

Order Now

Specification Test Results

Melting point 86-88°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR