N-Methylindole-6-carboxylic acid methyl ester ≥98%
Supplier: Matrix Scientific
Synonyms:
Methyl 1-methyl-1H-indole-6-carboxylate, Methyl 1-Methylindole-6-carboxylate
012577-1G
012577-5G
101802-898EA
26.15
USD
101802-898
101802-900
N-Methylindole-6-carboxylic acid methyl ester ≥98%
N-Methylindole-6-carboxylic acid methyl ester
Hazard: Irritant
Formula:
C₁₁H₁₁NO₂ Melting Pt: 90…93 °C |
MDL Number:
MFCD06245515 CAS Number: 1204-32-6 |
Specification Test Results
Melting point | 90-93°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...