4-Chloro-2-fluorophenylhydrazine hydrochloride ≥98%

Supplier: Matrix Scientific

Synonyms: (4-Chloro-2-fluorophenyl)hydrazine hydrochloride

012572-1G 012572-5G
101802-888EA 15.38 USD
101802-888 101802-890
4-Chloro-2-fluorophenylhydrazine hydrochloride ≥98%
4-Chloro-2-fluorophenylhydrazine hydrochloride

Hazard: Irritant

Formula: C₆H₆ClFN₂·ClH
Melting Pt: 234-235°C (dec)
MDL Number: MFCD06245512
CAS Number: 64172-78-7

Order Now

Specification Test Results

Melting point 234-235°C(d


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR