Terephthalamic acid ≥90%

Supplier: Matrix Scientific

Synonyms: Terephthalamidic Acid, Terephthalic acid monoamide

012552-1G 012552-5G
101802-836EA 32.31 USD
101802-836 101802-838
Terephthalamic acid ≥90%
Terephthalamic acid

Hazard: Irritant

Formula: C₈H₇NO₃
MW: 165.15 g/mol
Melting Pt: >340 °C
MDL Number: MFCD00025482
CAS Number: 6051-43-0

Order Now

Specification Test Results

Melting point >340°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR