2-Carbomethoxybenzylamine hydrochloride ≥90%

Supplier: Matrix Scientific

Synonyms: Methyl 2-(aminomethyl)benzoate hydrochloride

012548-1G 012548-5G
101802-822EA 1244.62 USD
101802-822 101802-824
2-Carbomethoxybenzylamine hydrochloride ≥90%
2-Carbomethoxybenzylamine hydrochloride

Hazard: Irritant/Stored under Argon

Formula: C₉H₁₂ClNO₂
Melting Pt: 170…172 °C (dec.)
MDL Number: MFCD06245496
CAS Number: 849020-92-4

Order Now

Specification Test Results

Melting point 170-172°C(d


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR