3-Fluoropiperidine hydrochloride ≥98%

Supplier: Matrix Scientific

007652-250MG 007652-1G
101793-182EA 83.08 USD
101793-182 101793-184
3-Fluoropiperidine hydrochloride ≥98%
3-Fluoropiperidine hydrochloride

Hazard: Irritant/Air Sensitive

Formula: C₅H₁₀FN·HCl
MW: 139.60 g/mol
Melting Pt: ∼ 227 °C
MDL Number: MFCD03452799
CAS Number: 737000-77-0

Order Now

Specification Test Results

Melting point ca 227°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR