2,4,5,6-Tetrachloro-m-xylylenediamine ≥97%
Supplier: Matrix Scientific
007646-1G
007646-5G
101793-170EA
36.92
USD
101793-170
101793-172
2,4,5,6-Tetrachloro-m-xylylenediamine ≥97%
2,4,5,6-Tetrachloro-m-xylylenediamine
Hazard: Irritant
Formula:
C₈H₈Cl₄N₂ Melting Pt: 133 °C |
MDL Number:
MFCD00025573 CAS Number: 16969-06-5 |
Specification Test Results
Melting point | 133°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...