3-Nitrobenzylamine hydrochloride ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Nitrobenzylammonium hydrochloride

007634-5G 007634-25G
101793-144EA 44.62 USD
101793-144 101793-146
3-Nitrobenzylamine hydrochloride ≥97%
3-Nitrobenzylamine hydrochloride

Hazard: Irritant/Hygroscopic/Store in Freezer

Formula: C₇H₉ClN₂O₂
MW: 188.62 g/mol
Melting Pt: 229…231 °C
MDL Number: MFCD00012858
CAS Number: 26177-43-5

Order Now

Specification Test Results

Melting point 229-231°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR