2-Naphthalenemethanol ≥98%

Supplier: Matrix Scientific

007631-5G 007631-25G
101793-140EA 36.92 USD
101793-140 101793-142
2-Naphthalenemethanol ≥98%
2-Naphthalenemethanol

Hazard: Irritant

Formula: C₁₁H₁₀O
MW: 158.2000 g/mol
Melting Pt: 80 °C
MDL Number: MFCD00004124
CAS Number: 1592-38-7

Order Now

Specification Test Results

Melting point 80°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR