6-Methoxy-m-toluic acid ≥98%

Supplier: Matrix Scientific

007614-1G 007614-5G
101793-116EA 53.85 USD
101793-116 101793-118
6-Methoxy-m-toluic acid ≥98%
6-Methoxy-m-toluic acid

Hazard: Irritant

Formula: C₉H₁₀O₃
Melting Pt: 69 °C
MDL Number: MFCD00045849
CAS Number: 25045-36-7

Order Now

Specification Test Results

Melting point 69°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR