3-(4-Methoxybenzoyl)propionic acid 98+%

Supplier: Matrix Scientific
Warning

Synonyms: 4-(4-Methoxyphenyl)-4-oxobutanoic acid

007612-5G 007612-25G
101793-108EA 36.92 USD
101793-108 101793-110
3-(4-Methoxybenzoyl)propionic acid 98+%
3-(4-Methoxybenzoyl)propionic acid

Purity: 98% CAS: 3153-44-4 MDL Number: MFCD00002795 Molecular Formula: C11H12O4 Molecular Weight: 208,22 Hazard: Irritant

Formula: C₁₁H₁₂O₄
MW: 208.21 g/mol
Melting Pt: 148…150 °C
MDL Number: MFCD00002795
CAS Number: 3153-44-4

Order Now

Specification Test Results

Melting point 148-150°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR