2,2'-Biimidazole ≥96%
Supplier: Matrix Scientific
Synonyms:
2,2'-Diimidazole, 2,2'-Bisimidazole, 1H,1'H-2,2'-Biimidazole
007513-1G
007513-5G
101792-956EA
80
USD
101792-956
101792-958
2,2'-Biimidazole ≥96%
2,2'-Biimidazole
Hazard: Corrosive
Formula:
C₆H₆N₄ MW: 134.14 g/mol Melting Pt: >350 °C |
MDL Number:
MFCD00047014 CAS Number: 492-98-8 |
Specification Test Results
Melting point | >350°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...