4-Iodobenzaldehyde ≥97%
Supplier: Matrix Scientific
007492-1G
007492-5G
007492-25G
101792-922EA
15.38
USD
101792-922
101792-924
102550-906
4-Iodobenzaldehyde ≥97%
4-Iodobenzaldehyde
Hazard: Irritant/Air Sensitive/Store in Freezer
Formula:
C₇H₅IO MW: 232.02 g/mol Melting Pt: 76…78 °C |
MDL Number:
MFCD00039576 CAS Number: 15164-44-0 |
Specification Test Results
Melting point | 76-78°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...