3-[4-(Methylthio)phenyl]propionic acid ≥97%
Supplier: Matrix Scientific
007480-1G
007480-5G
101792-898EA
98.46
USD
101792-898
101792-900
3-[4-(Methylthio)phenyl]propionic acid ≥97%
3-[4-(Methylthio)phenyl]propionic acid
Hazard: Irritant
Formula:
C₁₀H₁₂O₂S Melting Pt: 100…102 °C |
MDL Number:
MFCD03425696 CAS Number: 138485-81-1 |
Specification Test Results
Melting point | 100-102°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...