4-Methoxy-3-nitrobiphenyl ≥99%

Supplier: Matrix Scientific

Synonyms: 2-Nitro-4-phenylanisole

007477-5G 007477-25G
101792-886EA 53.85 USD
101792-886 101792-888
4-Methoxy-3-nitrobiphenyl ≥99%
4-Methoxy-3-nitrobiphenyl

Hazard: Irritant

Formula: C₁₃H₁₁NO₃
MW: 229.24 g/mol
Melting Pt: 90…91 °C
MDL Number: MFCD00191554
CAS Number: 15854-73-6

Order Now

Specification Test Results

Melting point 90-91°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR