3-(4-Isopropylphenyl)propionic acid ≥97%

Supplier: Matrix Scientific

007473-5G 007473-25G
101792-872EA 153.85 USD
101792-872 101792-874
3-(4-Isopropylphenyl)propionic acid ≥97%
3-(4-Isopropylphenyl)propionic acid

Hazard: Irritant

Formula: C₁₂H₁₆O₂
Melting Pt: 74…76 °C
MDL Number: MFCD01462077
CAS Number: 58420-21-6

Order Now

Specification Test Results

Melting point 74-76°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR