N-Isopropyl chloroacetamide ≥97%

Supplier: Matrix Scientific

Synonyms: N-Isopropyl-2-chloroacetamide

007471-1G 007471-5G
101792-864EA 52.31 USD
101792-864 101792-866
N-Isopropyl chloroacetamide ≥97%
N-Isopropyl chloroacetamide

Hazard: Irritant

Formula: C₅H₁₀ClNO
MW: 135.59 g/mol
Melting Pt: 58…60 °C
MDL Number: MFCD00068034
CAS Number: 2895-21-8

Order Now

Specification Test Results

Melting point 58-60°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR