3-(4-Iodophenyl)propionic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 3-(4-Iodophenyl)propanoic acid

007468-1G 007468-5G
101792-852EA 93.85 USD
101792-852 101792-854
3-(4-Iodophenyl)propionic acid ≥98%
3-(4-Iodophenyl)propionic acid

Hazard: Irritant

Formula: C₉H₉IO₂
MW: 276.08 g/mol
Melting Pt: 140…142 °C
MDL Number: MFCD00017597
CAS Number: 1643-29-4

Order Now

Specification Test Results

Melting point 140-142°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR