3-(2,4-Dimethylphenyl)propionic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(2,4-Dimethylphenyl)propanoic acid

007461-1G 007461-5G
101792-830EA 136.92 USD
101792-830 101792-832
3-(2,4-Dimethylphenyl)propionic acid ≥97%
3-(2,4-Dimethylphenyl)propionic acid

Hazard: Irritant

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Melting Pt: 106…108 °C
MDL Number: MFCD00796498
CAS Number: 1811-85-4

Order Now

Specification Test Results

Melting point 106-108°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR