3-(2,3-Dimethoxyphenyl)propionic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 3-(2,3-Dimethoxyphenyl)propanoic acid

007458-1G 007458-5G
101792-818EA 36.92 USD
101792-818 101792-820
3-(2,3-Dimethoxyphenyl)propionic acid ≥98%
3-(2,3-Dimethoxyphenyl)propionic acid

Hazard: Irritant

Formula: C₁₁H₁₄O₄
MW: 210.23 g/mol
Melting Pt: 58-60°C/1mm
MDL Number: MFCD00029821
CAS Number: 10538-48-4

Order Now

Specification Test Results

Melting point 58-60°C/1mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR