1,4-Diethoxytetrafluorobenzene

Supplier: Matrix Scientific

007427-5G 007427-25G
101792-734EA 56.92 USD
101792-734 101792-736
1,4-Diethoxytetrafluorobenzene
1,4-Diethoxytetrafluorobenzene

Hazard: Irritant

Formula: C₁₀H₁₀F₄O₂
MW: 238.18 g/mol
Melting Pt: 49.0…49.5 °C
MDL Number: MFCD00804629
CAS Number: 16251-00-6

Order Now

Specification Test Results

Melting point 49.0-49.5°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR