1-Bromo-3,5-dichloro-2,4,6-trifluorobenzene ≥97%

Supplier: Matrix Scientific

007421-5G 007421-25G
101792-712EA 249.23 USD
101792-712 101792-714
1-Bromo-3,5-dichloro-2,4,6-trifluorobenzene ≥97%
1-Bromo-3,5-dichloro-2,4,6-trifluorobenzene

Hazard: Irritant

Formula: C₆BrCl₂F₃
Melting Pt: 58…60 °C
MDL Number: MFCD03424469
CAS Number: 24812-13-3

Order Now

Specification Test Results

Melting point 58-60°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR