5-(Piperidin-4-yl)-3-[3-(trifluoromethyl)phenyl]-1,2,4-oxadiazole

Supplier: Matrix Scientific

Synonyms: 4-{3-[3-(Trifluoromethyl)phenyl]-1,2,4-oxadiazol-5-yl}piperidine

007360-500MG 007360-1G
101792-548EA 207.69 USD
101792-548 101792-550
5-(Piperidin-4-yl)-3-[3-(trifluoromethyl)phenyl]-1,2,4-oxadiazole
5-(Piperidin-4-yl)-3-[3-(trifluoromethyl)phenyl]-1,2,4-oxadiazole

Hazard: Irritant

Formula: C₁₄H₁₄F₃N₃O
MW: 297.28 g/mol
Melting Pt: 69…71 °C
MDL Number: MFCD03424498
CAS Number: 808764-45-6

Order Now

Specification Test Results

Melting point 69-71°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR