2-Amino-6-fluorobenzamide
Supplier: Matrix Scientific
007344-1G
007344-5G
007344-25G
101792-508EA
23.08
USD
101792-508
101792-510
101792-512
2-Amino-6-fluorobenzamide
2-Amino-6-fluorobenzamide
Hazard: Irritant
Formula:
C₇H₇FN₂O Melting Pt: 115…118 °C |
MDL Number:
MFCD00816881 CAS Number: 115643-59-9 |
Specification Test Results
Melting point | 115-118°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...