Adamantane-1-carboxamidine hydrochloride

Supplier: Matrix Scientific

007343-1G 007343-5G
101792-504EA 92.31 USD
101792-504 101792-506
Adamantane-1-carboxamidine hydrochloride
Adamantane-1-carboxamidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₉ClN₂
Melting Pt: 255 °C
MDL Number: MFCD00167793
CAS Number: 50417-14-6

Order Now

Specification Test Results

Melting point 255°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR